Documenti fondamentali
BR462231
BRAND® rubber sleeve for filter crucibles
O.D. 49 mm
Sinonimo/i:
Filter, Rubber sleeve
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.72
Materiali
EPDM rubber
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
BRAND 462231
D.E.
49 mm
Descrizione generale
The BRAND® Rubber Sleeve for Filter Crucibles is made from high-quality EPDM rubber, designed to provide a secure and reliable fit for various filter crucibles according to the identification code. This durable sleeve enhances the functionality of filter crucibles by ensuring a tight seal, preventing leaks and contamination during filtration processes.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)