BR452201
BRAND® Inoculation loop with needle, disposable
volume 1 μL, white, sterile; γ-irradiated
Sinonimo/i:
Inoculation loops
About This Item
Codice UNSPSC:
41161502
NACRES:
NB.22
Prodotti consigliati
Materiali
polystyrene
white
Sterilità
sterile; γ-irradiated
Confezionamento
pack of 1000 ea
pkg of 1000 ea (50 bags of 20)
Produttore/marchio commerciale
BRAND 452201
Volume
1 μL
Descrizione generale
For inoculation of nutrient media. Available either with loop at one end and needle at the other or loops at both ends. High flexibility permits gentle streaking without damaging the nutrient surface. γ-ray sterilized.
Caratteristiche e vantaggi
- Sterile (SAL 10-6)
- High flexibility for agar-saving smears
- Usable at both ends
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)