BR32805
BRAND® USP BLAUBRAND® class A measuring cylinder, tall form
volume 5 mL
Sinonimo/i:
BLAUBRAND®, Graduated Cylinders, USP
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
Caratteristiche
DE-M marking
Produttore/marchio commerciale
BRAND 32805
Volume
5 mL
Descrizione generale
The BRAND® USP BLAUBRAND® class A measuring cylinder in tall form is crafted from high-quality Boro 3.3 glass, ensuring durability and precision. It complies with DIN EN ISO 4788 standards and is calibrated to contain (TC, In), making it suitable for accurate liquid measurement. The design features a spout for easy pouring and a hexagonal glass base for stability. Each measuring cylinder comes with a USP batch certificate. This measuring cylinder is an essential tool for laboratories requiring high accuracy and reliability in liquid measurement.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)