BR28305
BRAND® pipette tray
PVC, L × W × H 420 mm × 300 mm × 30 mm
Sinonimo/i:
Pipette Trays, Trays
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.51
Prodotti consigliati
Materiali
PVC
Produttore/marchio commerciale
BRAND 28305
Lungh. × Largh. × Alt.
420 mm × 300 mm × 30 mm
Descrizione generale
The BRAND® Pipette Tray is made from durable PVC and is designed as an efficient drawer organizer for graduated and bulb pipettes.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
BR28305-1EA | 4061831855531 |