BR150902
BRAND® watch glass
PTFE, diam. 30 mm
Sinonimo/i:
Dishes, PTFE, Watch glasses
About This Item
Codice UNSPSC:
41121801
NACRES:
NB.43
Prodotti consigliati
Materiali
PTFE
Produttore/marchio commerciale
BRAND 150902
Diametro
30 mm
Descrizione generale
The BRAND® watch glass is made from high-quality PTFE, making it ideal for covering PTFE beakers. Its excellent chemical resistance ensures durability and reliability when handling a wide range of substances. This watch glass provides a secure cover to prevent contamination and evaporation during experiments. Its lightweight and non-reactive properties make it a valuable accessory in laboratory settings, enhancing the efficiency of various applications. This versatile tool is essential for maintaining the integrity of samples and solutions in the lab.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR150902-1EA | 4061831854053 |