BR114817
BRAND® PP graduated centrifuge tube, screw cap
volume 15 mL, without base, non-sterile
Sinonimo/i:
Centrifuge tubes, Tubes
About This Item
Codice UNSPSC:
41121703
NACRES:
NB.11
Prodotti consigliati
Materiali
polypropylene
screw top cap
translucent tube
Sterilità
non-sterile
Caratteristiche
RCF 3,000 × g
without base
Confezionamento
pack of 750
Produttore/marchio commerciale
BRAND 114817
Volume
15 mL
Categorie correlate
Descrizione generale
The BRAND® PP Graduated Centrifuge Tube with Screw Cap is made from high-quality polypropylene (PP) and features a translucent design for easy visibility of contents. This tube is engineered to withstand relative centrifugal forces (RCF) of up to 3000, making it suitable for various laboratory applications. It comes graduated, allowing for precise measurements, and is available in both non-sterile and gamma-ray sterilized options in 15 ml (graduated up to 13 ml) or 50 ml capacities. The screw cap, made from polyethylene (PE) and equipped with a sealing cone, ensures a secure closure to prevent leaks. This centrifuge tube offers high clarity and reliability, making it an essential tool for researchers and technicians in the lab.
Caratteristiche e vantaggi
- BRAND® graduated centrifuge tube is the ideal solution for all your centrifuge needs.
- Made from highly transparent polypropylene (PP) for enhanced sample visibility.
- Bio-inert PP offers optimal cell and protein recovery.
- Includes a screw cap with a plug for preventing sample loss.
- Tubes are tested to withstand RCF up to 3000.
- γ-ray sterilized.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)