BAH500250310
SP Bel-Art® Glove Box Bellows Type Gloves for 8 in. Glove Ports
latex, size 10 large
Sinonimo/i:
glove box
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.43
Prodotti consigliati
Materiali
latex
Misura
10 large
Caratteristiche
to hold Bellows and Gloves
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art H50025-0310
Descrizione generale
Made from natural latex with a removable hand, the sleeves fit 20cm (8") circular ports and 18 x 25cm (7 x 10") oval ports. Use with H50025-0320 Clamping Rings to hold gloves in place.
Caratteristiche e vantaggi
- Forms a leak-proof unit with O-ring construction integral with the box
- Bellows allows cooling air circulation
- When the glove and sleeve are separated, the sleeve remains attached to the box
- Glove sizes are interchangeable, and almost any standard laboratory, surgical or disposable glove can be used
- Length of the extended sleeve and glove is approximately 76cm (30")
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)