BAH131761014
SP Bel-Art® utility bags
width × H × thickness 10 in. (254 mm) × 14 in. (356 mm) × 1.5 mil (0.038 mm), clear, pack of 100 ea
Sinonimo/i:
clear bag, reusable bag, sample bag
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.35
Materiali
clear
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
Bel-Art H13176-1014
width × H × thickness
10 in. (254 mm) × 14 in. (356 mm) × 1.5 mil (0.038 mm)
Descrizione generale
Reusable, transparent bags are ideal for shipping or storage and are especially useful as sample bags.
Caratteristiche e vantaggi
- FDA grade polyethylene, safe for food, drug and cosmetic use
- Latex free
- Not Autoclavable
- H13176 lightweight bags are 0.038 mm (1.5 mil) thick, H13178 heavyweight bags are 0.1016 mm (4 mil) thick
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)