Documenti fondamentali
BAF430000001
Shelf set for Scienceware® Grande desiccator
acrylic, perforated, pack of 2 ea
Sinonimo/i:
Scienceware, dry-box desiccator cabinet, low humidity
About This Item
Codice UNSPSC:
41104510
NACRES:
NB.43
Caratteristiche
acrylic, perforated
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
Bel-Art F43000-0001
Descrizione generale
Grande Desiccator can hold up to seven shelves spaced 2 inches apart, pack of 2 (two are included with desiccator).
  • Pull-out, perforated acrylic shelves allow maximum air circulation and have built-in stops to prevent accidental tipping; maximum weight per shelf: 10 lbs (4.5 kg)
  • Shelves are removable to allow use as a single, spacious chamber for specific storage needs
  • Dimensions: 50.8 W x 53 D x 6.7 cm H (20 x 20 7/8 x 2 5/8 in.
The desiccant used in the cartridge is: Z163414
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)