BAF369750600
SP Bel-Art® Sterileware™ Powder Probe Samplers
white high-density polyethylene, capacity 125 mL (4 oz), L 61.5 cm (24.25 in.), sterile, pack of 20 ea (individually wrapped)
Sinonimo/i:
sampler
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.35
Prodotti consigliati
Materiali
white high-density polyethylene
Sterilità
sterile
Confezionamento
pack of 20 ea (individually wrapped)
Produttore/marchio commerciale
Bel-Art F36975-0600
Lunghezza
61.5 cm (24.25 in.)
Capacità
125 mL (4 oz)
Descrizione generale
Ideal for taking a cross sectional sample through a bulk powder, this probe is available in two convenient lengths. Used by pharmaceutical, food, cosmetic and chemical companies.
Caratteristiche e vantaggi
- End tip contains free-flowing powders within the probe
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) probe is heat sealed in individual polyethylene bag
- Gamma irradiated sterile (SAL 10^-6)Ready for one time use and disposal
- Lot stamped for accurate tracing Note: Sterilized products are not returnable
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)