Documenti fondamentali
BAF169570000
SP Bel-Art® Cone Style Acid/Solvent Bottle Carrier
to hold, one 2.5 Liter Bottle (5 pints)
Sinonimo/i:
solvent bottle carrier
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.43
Materiali
conical
Caratteristiche
to hold one 2.5 Liter Bottle (5 pints)
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F16957-0000
diam. × alt.
7.125 in. (180 mm) × 13.5 in. (350 mm)
Descrizione generale
Polyethylene carriers with epoxy coated, heavy-gauge steel wire bails have wide ribs in both the sides and on the bottom to support and cushion bottles.
Caratteristiche e vantaggi
  • Cone style lid has a 16mm (⅝”) opening in the top for insertion of tubing; simplifies the transfer of liquids without pouring
  • Truncated style lid permits pouring without the removal of the lid
  • Designed to hold standard US reagent bottles
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)