BAF131930102
Clavies® cover for biohazard bag holder
bench-top
About This Item
Codice UNSPSC:
24111500
NACRES:
NB.35
Prodotti consigliati
Materiali
polypropylene
Caratteristiche
autoclavable
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F13193-0102
Misura
203x241 mm
Categorie correlate
Descrizione generale
Dishwasher and autoclave safe, the reusable Clavies® Biohazard Bag Holder Cover opens quickly and easily by pressing the built-in release tab – no batteries required! The cover fits both the Medium and Large sizes of the Clavies Biohazard Bag Holders.
- Opens with a single touch; sanitary, reusable
- Can be installed in seconds
- Improves the sanitation of these biohazardous waste containers
- Center panel of the lid opens to an upright position and reveals a large 20.3 x 24.1cm (8 x 9 1/2 in.) opening through which wastes can be deposited
Note legali
Clavies is a registered trademark of SP Scienceware, a division of SP
Prodotti correlati
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)