BAF118590010
SP Bel-Art® jerrican with spigot
capacity 5 L (1.25 gal), high-density polyethylene
Sinonimo/i:
jerrycan, portable dispensor
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.43
Prodotti consigliati
Materiali
high-density polyethylene
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F11859-0010
Capacità
5 L (1.25 gal)
Descrizione generale
Space-saving jerricans have a smooth turning spigot that opens and closes with half a turn for easy convenient dispensing from a stationary position. Made of high-density polyethylene, they have a unique 20cm (8") long spout with a built-in air vent. The 5 gallon jerrican has a 2.5cm (1") I.D. pouring spout; other sizes have 1.9cm (¾") I.D. spouts.
Caratteristiche e vantaggi
- Great for dispensing without moving
- Spout retracts into the threaded collar to conserve space
- Collar is sealed by leakproof screw cap
- Extra hand grip on the bottom gives positive control when pouring
- Extra headspace for expansion of contents
- Graduated in U.S. gallons and liters
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)