ACE6893015
Ace receiving flask for rotary evaporators, graduated, round bottom
size 500 mL
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass flask
round bottom
single-neck
Giunto
joint 35/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Ace Glass 6893-15
Misura
500 mL
Categorie correlate
Descrizione generale
Graduated, round bottom receiving flask for use with all rotary evaporators. 35/20 spherical outer joint will also mate to 35/25 ball joints. 250 mL and 500 mL size graduated in 10 mL increments, 1000 mL and 2000 mL in 100 mL increments.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
ACE6893015-1EA | 4061832346137 |