Skip to Content
Merck
All Photos(1)

Key Documents

R0775

Sigma-Aldrich

RANK/Fc Chimera from mouse

≥95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture

Synonym(s):

Osteoclast differentiation receptor, ODFR, Receptor activator of NF-κB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

mouse

Quality Level

recombinant

expressed in NSO cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

10.12 μg/mL EC50

mol wt

60 kDa

packaging

pkg of 1 mg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

Application

RANK is the essential signaling receptor for osteoclast differentiation factor in osteoclastogenesis. The biological functions mediated by RANK include activation of NF-ΚB and c-jun N-terminal kinase, enhancement of T cell growth and dendritic cell function, induction of osteoclastogenesis and lymph node organogenesis.

Biochem/physiol Actions

TRANCE (TNF-related activation-induced cytokines, also known as RANK ligand, osteoprotegerin ligand and osteoclast differentiation factor) is the ligand for RANK. The biological functions mediated through RANK include activation of NF-κB and c-jun N-terminal kinases, enhancement of T cell growth and dentritic cell function, induction of osteoclastogenesis, and lymph node genesis. Soluble RANK is able to block TRANCE-induced biological activity.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

The biological activity is measured by its ability to inhibit TRANCE-induced osteoclast differentiation in mouse splenocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Nakagawa et al.
Biochemical and biophysical research communications, 253(2), 395-400 (1999-01-08)
Osteoclast differentiation factor (ODF) is a ligand for osteoclastogenesis-inhibitory factor/osteoprotegerin (OCIF/OPG), and mediates an essential signal for osteoclastogenesis. Soluble-form ODF binds directly to osteoclast progenitors, suggesting the presence of a membrane-bound receptor for ODF (ODFR) on the cells. To understand
D M Anderson et al.
Nature, 390(6656), 175-179 (1997-11-21)
Dendritic cells are rare haematopoietic cells that reside in a number of organs and tissues. By capturing, processing and presenting antigens to T cells, dendritic cells are essential for immune surveillance and the regulation of specific immunity. Several members of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service