Skip to Content
Merck
All Photos(1)

Key Documents

05-1516

Sigma-Aldrich

Anti-hnRNP U Antibody, clone 3G6

clone 3G6, from mouse

Synonym(s):

Scaffold attachment factor A, heterogeneous nuclear ribonucleoprotein U, heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A), hnRNP U, hnRNP U protein, p120 nuclear protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G6, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HNRNPU(3192)

General description

Heterogeneous nuclear ribonucleoproteins (hnRNPs) are thought to be involved in pre-mRNA processing. However, its role in the regulation of gene expression is as yet poorly understood. Proteins of the heterogeneous nuclear ribonucleoparticles (hnRNP) family form a structurally diverse group of RNA binding proteins implicated in various functions. Recently, hnRNP proteins have been shown to hinder communication between factors bound to different splice sites. Conversely, several reports have described a positive role for some hnRNP proteins in pre-mRNA splicing. hnRNP-U, also termed scaffold attachment factor A (SAF-A), binds to pre-mRNA and nuclear matrix/scaffold attachment region DNA elements.

Specificity

This antibody recognizes hnRNP U.

Immunogen

Epitope: Unknown
Recombinant protein corresponding to human hnRNP U.

Application

Immunoprecipitation
Reported by external laboratory using a represenative lot of this antibody
Immunocytochemistry
Reported by external laboratory using a represenative lot of this antibody
ELISA
Reported by external laboratory using a represenative lot of this antibody
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Apoptosis - Additional
Use Anti-hnRNP U Antibody, clone 3G6 (mouse monoclonal antibody) validated in WB, IP, ICC, ELISA to detect hnRNP U also known as heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A).

Quality

Evaluated by Western Blot in K562 cell lysate.

Western Blot Analysis: 0.01 µg/ml of this antibody detected hnRNP U on 10 µg of K562 cell lysate.

Target description

Calculated molecular weight is 90 kDa However protein is usually observed at ~120 kDa (Dreyfuss G, Kim VN, Kataoka N. Nat Rev Mol Cell Biol. 2002 Mar;3(3):195-205.)

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L Baeza et al.
The Journal of clinical investigation, 85(5), 1516-1521 (1990-05-01)
We have previously reported purification of three forms of histamine-releasing factors (HRFs) from mixtures of streptokinase-streptodornase stimulated human mononuclear cells and platelets with apparent molecular masses of 10-12, 15-17, and 40-41 kD (1989. J. Clin. Invest. 83:1204-1210). We have also
Adam Buckle et al.
Human molecular genetics, 27(19), 3434-3448 (2018-07-15)
Complex diseases, such as diabetes, are influenced by comprehensive transcriptional networks. Genome-wide association studies have revealed that variants located in regulatory elements for pancreatic transcription factors are linked to diabetes, including those functionally linked to the paired box transcription factor
Roi Isaac et al.
iScience, 25(11), 105270-105270 (2022-10-29)
The seven-transmembrane superfamily member 3 protein (TM7SF3) is a p53-regulated homeostatic factor that attenuates cellular stress and the unfolded protein response. Here we show that TM7SF3 localizes to nuclear speckles; eukaryotic nuclear bodies enriched in splicing factors. This unexpected location

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service