Merck
58256
Edelstahl-HPLC-Verschraubungen
nut for 1/16 in. tubing, pkg of 10 ea
About This Item
UNSPSC-Code:
41115700
Materialien
stainless steel nut
Verpackung
pkg of 10 ea
Hersteller/Markenname
Rheodyne 6000-263
Fitting
nut for 1/16 in. tubing
10-32 Rheodyne for 1/16 in. O.D.
Allgemeine Beschreibung
The long Rheodyne stainless steel nut of the Stainless Steel HPLC Fitting has a longer hex portion for easier wrench access.
Empfohlene Produkte
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)