Merck
Z659118
Bandelin SONOPULS ultrasonic homogenizer standard and booster horns
SH 213G booster horn for HD 2200
Synonym(e):
ultrasonic booster
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.74
Materialien
titanium
Hersteller/Markenname
Bandelin
Verfügbarkeit
available only in EU
Durchmesser
13 mm
Allgemeine Beschreibung
Standard and booster horns (Ti-6AI-4V) are furnished with a thread for replaceable probes. With an exterior thread to connect various vessels.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)