Merck
TMO312760-0040
Nalgene® closure for dropper Bottle
green polypropylene, diam. 16 mm, thread 15-415, case of 2000 ea
Synonym(e):
dropping bottle closure, dropping bottle seal
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
green polypropylene
Verpackung
case of 2000 ea
Hersteller/Markenname
Nalgene 312760-0040
Durchmesser
16 mm
Gewinde
15-415
Allgemeine Beschreibung
Order Thermo Scientific Nalgene Closures for Dropper Bottles to complete the Nalgene Dropper Bottle System. Closures fit dropper bottles 312750 and 312751 and are available in multiple colors to help identify reagents.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)