Merck
TMO2403-1000
Nalgene® phthalate-free wash bottle
FEP, bottle volume 1000 mL, unlabelled, case of 4 ea
Synonym(e):
fluid diepenser, plastic wash bottle
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
FEP
Verpackung
case of 4 ea
Hersteller/Markenname
Nalgene 2403-1000
Flaschenvolumen
1000 mL
Allgemeine Beschreibung
Ensure contamination-free dispensing with Thermo Scientific Nalgene Wash Bottles made with PTFE FEP, useful for trace element work. These phthalate-free bottles offer excellent chemical resistance and are molded entirely of fluoropolymers. Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. The precision-molded tip provides a straight, steady stream and ultimate control without sputtering.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)