Merck
OH30059253
Ohaus® Reference Electrode
STREF1
About This Item
UNSPSC-Code:
39121436
Verpackung
pack of 1 ea
Hersteller/Markenname
Ohaus® 30059253
Allgemeine Beschreibung
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Anwendung
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Leistungsmerkmale und Vorteile
Can be used in conjunction with all Starter bench and portable meters
Rechtliche Hinweise
Ohaus is a registered trademark of Ohaus Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)