Merck
MTCP3801QW
MTC Bio Pure.Amp PCR 8 Tube Strips
new
capacity 0.1 mL, white tube, separate, optical clear flat cap
Synonym(e):
PCR sample storage tubes, plastic tube, sample storage tube
About This Item
UNSPSC-Code:
41121800
Materialien
white tube
Verpackung
pack of 120 ea
Hersteller/Markenname
MTC Bio P3801-QW
Kapazität
0.1 mL
Allgemeine Beschreibung
0.1ml qPCR 8-Strip (With SEPARATE Optical Strip Caps), WHITE 120/pk
Leistungsmerkmale und Vorteile
  • RNase, DNase, DNA, and PCR inhibitor free
  • 0.2mL or Low Profile 0.1mL options
  • Ultra-thin, uniform wall thickness
  • White and frosted tubes for enhanced qPCR results
Rechtliche Hinweise
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio