Merck
GBL845233
Grace Bio-Labs ProCrystal protein crystallization cover
uncut, plastic liner
Synonym(e):
adhesive crystallization cover, precut paper frame crystallization cover
About This Item
UNSPSC-Code:
41122410
NACRES:
NB.15
Materialien
plastic film
Sterilität
non-sterile
Verpackung
pack of 5 ea
Hersteller/Markenname
Grace Bio-Labs 845233
B × L
78 mm × 120 mm
Allgemeine Beschreibung
ProCrystal protein crystallization covers are designed for high throughput protein x-ray crystallography in 96-well plates. The ProCrystal material is extremely hydrophobic to maintain drop footprint and segregation, even with protocols using MPD, glycerol or detergents. Grace Bio-Labs has collaborated with leading crystallographers to develop our ProCrystal covers using materials with the highest optical quality and stringent manufacturing processes to ensure clean, particle-free covers.
Features Include:
  • TTP Labtech Mosquito compatible
  • Extremely hydrophobic
  • Ready-to-use, dust-free
  • UV compatible
  • No noticeable x-ray diffraction
  • Accommodates up to 3 protein drops per well
  • Two formats: pre-cut or uncut
ProCrystal protein crystallization covers are extremely hydrophobic and UV compatible. Covers accommodate up to 3 protein drops per well, and are available as frame or frameless, with either paper or plastic liners, and in two cut formats: pre-cut or uncut. Our protein crystallization covers are compatible with SBS-compliant microtiter plates and TTP Mosquito Instruments. The ProCrystal cover is also UV compatible with the highest optical quality available. The stringent manufacturing process and quality control ensure clean, particle-free material shipped ready to use. ProCrystal protein crystallization covers are available in an uncut, or pre-cut format that allows removal of covers from individual wells at different times.
Rechtliche Hinweise
ProCrystal is a trademark of Grace Bio-Labs, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
GBL845233-5EA4061832401393