Merck
Z764531
2mag bioMIXcontrol S control unit
AC/DC input 230 V AC, UK plug
Synonym(e):
induction stirrer, magnetic induction stir plate, magnetic wear-free stirrer, stirrer
About This Item
UNSPSC-Code:
41103800
NACRES:
NB.72
Materialien
stainless steel
Verpackung
pack of 1 ea
Hersteller/Markenname
2mag 98500
Parameter
5-250 rpm speed
B × T × H
215 mm × 120 mm × 55 mm
Plattengröße × B × L
× 200 mm × 250 mm
Gewicht
1.4 kg
Ausgabe
4-40 W (stirring power)
Allgemeine Beschreibung
  • Efficient control unit, for 2mag bioMIXdrive series, developed for the hard daily lab use and uncompromising warming-free continuous operation
  • Control unit for the connection of 1 stirring drive bioMIXdrive 1, 2, 3, 4, or 6 with 6 watts/stirring point, without any own-generated heating effect, extended speed range from 5 to 250 rpm, jerk-free stirring, even and smooth stirring also at slow speeds
  • Power setting (10-step) for high power with regard to large and viscous stirring amounts and reduced power for the warming-free continuous operation, e.g. in CO2-incubators
  • Identical to bioMIXcontrol, but with stackable housing and vertical control panel
  • Digital display for speed- and power setting, SoftStart for reliable and safe accelerating for stirring bar, QuickSet for quick setting of start- and maximum speed, storage of last working status when turning off, firm stainless steel housing, and easy to clean
Erforderlich, aber nicht bereitgestellt
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)