Merck
Z764124
KDS Adagio syringe pump software
Works for all KDS Legato pumps
Synonym(e):
dosing pump, infusion pump, pump infusion
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Verpackung
pack of 1 ea
Hersteller/Markenname
KD Scientific 788000
Allgemeine Beschreibung
Features:
  • Low Cost Simple Installation
  • Flow Evolution Graph
  • Import & Export Programs
  • Quick & Easy Manual Pump Control
  • Monitor One or More Pumps
  • Program Data Logging
Quick & Easy Manual Pump Control:
The manual pump control tool allows easy direct control of the pump, and pump commands can be entered directly into the log.
Multiple programs can be opened at the same time. The programs progression is tracked, and can be stored in a file for later access.
Systemvoraussetzungen
System Requirements:
  • 1 GHz Pentium processor or higher
  • 512 MB of RAM (1 GB recommended)
  • Windows 7 or Windows 10
  • Free RS232 or USB 2.0 ports (depending on the quantity, model and connectivity of the controlled pumps)
  • Daisy chained pumps require a single port.
  • Direct PC to Pump connections require one free port per pump.
  • Microsoft Excel 97 or higher
  • USB must remain inserted in PC for proper function of software.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)