Z569666
KDS syringe multi-rack for Legato™ series pumps
microliter
Synonym(e):
dosing pump syringe rack, infusion pump syringe rack
Alle Fotos(2)
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.01
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
KD Scientific 78-8302
Allgemeine Beschreibung
Accommodates up to 4 x 0.5 μL to 10 mL syringes. Works with Legato 200, 210 and 210P.
Rechtliche Hinweise
Legato is a trademark of KD Scientific
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)