Z742263
DURAN® GL 56 Bottle Tag
orange silicone
Synonym(e):
292435626
Alle Fotos(2)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Empfohlene Produkte
Materialien
orange silicone
Verpackung
pack of 20 ea
Hersteller/Markenname
DURAN® 292435626
Verwandte Kategorien
Allgemeine Beschreibung
Bottle tags can be used on their own for color identification or to secure the protective light sheild around the bottle. The GL 56 bottle tags are available in four colors (orange, yellow, blue and purple).
Leistungsmerkmale und Vorteile
- Manufactured from silicone rubber
- The Bottle Tags can be used for colour identification or to secure the protective Light Shield around the bottle
- The elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
- Feature a graspable tab that makes them easy to remove with bare or gloved hands
- Chemically robust, heat resistant and reusable
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z742263-20EA | 4061832802398 |