Z516198
Metals / metal hydrides test kit
for determining reagent purities
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.43
Empfohlene Produkte
Verwandte Kategorien
Allgemeine Beschreibung
This unique testing kit provides a convenient and highly accurate method for determining reagent purities. Hydrogen gas is produced when active metals and metal hydrides (including boron hydrides) react with suitable hydrolytic solvents. The hydrogen gas displaces the water in the graduated burette which allows for quantitative measurement.
The metals and metal hydrides which can be analyzed include:
Please refer to Aldrich Technical Information Bulletin AL-123 for procedural details of the testing process. This bulletin is downloadable from the Technical Reference section on our website at sigma-aldrich.com, or by calling your local Technical Service group. For replacement parts, visit sigma-aldrich.com/glass.
The metals and metal hydrides which can be analyzed include:
- Sodium and lithium as dispersions in mineral oil
- Borane-tetrahydrofuran and borane-methyl sulfide complexes
- 9-BBN
- K-Selectride
- Lithium aluminum hydride
- Diisobutylaluminum hydride
Please refer to Aldrich Technical Information Bulletin AL-123 for procedural details of the testing process. This bulletin is downloadable from the Technical Reference section on our website at sigma-aldrich.com, or by calling your local Technical Service group. For replacement parts, visit sigma-aldrich.com/glass.
Ersatzteil
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)