Merck
Z341878
IKA® universal attachments for KS 130/260 basic and control shakers
AS 130.1 universal attachment
About This Item
UNSPSC-Code:
41103811
NACRES:
NB.73
Verpackung
pkg of 1 ea
Hersteller/Markenname
IKA 8017300
Allgemeine Beschreibung
Note: This attachment can be adapted to all kinds of vessels by means of universal, infinitely variable clamping rolls.
Rechtliche Hinweise
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)