Z319260
Drying tube sublimation insert for Büchi® models B-580 & B-585 ovens
borosilicate glass, pkg of 1 ea
Alle Fotos(1)
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.43
Empfohlene Produkte
Materialien
borosilicate glass
Verpackung
pkg of 1 ea
Hersteller/Markenname
Büchi 037133
Allgemeine Beschreibung
As an alternative use, the Buchi B-585 glass drying oven (Z563080 or Z616818) has a sublimation drying tube insert where substances can be physically cleansed via sublimation. A solid matter passes directly into its gaseous state and is resublimated on a cold surface. The sublimation accessory is inserted into the drying oven and cooled with water or another cooling medium.
Rechtliche Hinweise
Büchi is a registered trademark of Büchi Labortechnik AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z319260-1EA | 4061832770833 |