Z265063
Butane fuel Micro Torch
MT-51 handheld torch
Synonym(e):
butane torch, butane torches, mini torch
Alle Fotos(1)
About This Item
UNSPSC-Code:
40101834
NACRES:
NB.51
Empfohlene Produkte
Allgemeine Beschreibung
Generate a 2,400°F (1,300°C) pinpoint flame for a wide variety of torch applications. Units are self-igniting and have an adjustable flame. Unit shipped empty, order butane separately.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z265063-1EA | 4061833293966 |