Z260266
FORTUNA® Universal Optifix® HF dispenser
HF model, volume 2-10 mL
Synonym(e):
HF Dispenser
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Empfohlene Produkte
Leistungsmerkmale
subdivision 0.2 mL
Hersteller/Markenname
Fortuna Graf 101.106-38
Messbereich Volumen
2-10 mL
Volumen
2-10 mL
Fitting
GL 40
GL 45
Verwandte Kategorien
Allgemeine Beschreibung
For all liquids that can be handled with the Optifix dispensers and for hydrofluoric acid (HF)
Features:
Supplied:
FORTUNA Universal Optifix HF dispenser, with 2 PTFE adapters GL40 + GL45 & 1 PTFE intake tube
Features:
- Dispensing cylinder made of PTFE
- Massive PTFE encased dispensing piston
- FIX adapter made of PTFE
- Ergonomic finger protection cap
- Fast and easy volume setting with additional fine adjustment
- Valve block made of PTFE, with additional safety valve in the ventilation tube, no metal springs in the valves - no corrosion
Supplied:
FORTUNA Universal Optifix HF dispenser, with 2 PTFE adapters GL40 + GL45 & 1 PTFE intake tube
Rechtliche Hinweise
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z260266-1EA | 4061833578322 |