Z260231
FORTUNA® Optifix® Safety bottle-top dispenser
Safety model, volume 10-50 mL
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Empfohlene Produkte
Leistungsmerkmale
subdivision 1.0 mL
Hersteller/Markenname
Fortuna Graf 101.107-48
Messbereich Volumen
10-50 mL
Volumen
10-50 mL
Fitting
GL 40
GL 45
Verwandte Kategorien
Allgemeine Beschreibung
For dispensing strong acids and aggressive chemicals
Features:
Features:
- PTFE encased dispensing piston with sealing lip at its end
- FIX adapter made of PTFE
- Valve block made of PTFE, no metal springs in the valve - no corrosion
- Visible dispensing process by the use of a dispensing cylinder made of glass, coated with plastic-protection against breakage
- Ergonmic finger protection cap
- Fast and easy volume setting with additional fine adjustment
Rechtliche Hinweise
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z260231-1EA | 4061833568972 |