Merck
Z255696
Sigma-Aldrich
Kontes® NMR tube-sealing manifold
Use with 5mm × 9in. L NMR tubes
Synonym(e):
NMR-Röhrchen
About This Item
UNSPSC-Code:
41121705
NACRES:
SB.32
Hersteller/Markenname
Kimble®
Allgemeine Beschreibung
Single port design with HI-VAC® control valve and aluminum shield for heat deflection. Permits evacuation and flame tip-off of 5mm diam. NMR tube without additional sealing to other glass apparatus. To use, insert NMR tube through the open top cap, place Viton® O-ring over open end of tube, insert tube into the BEVEL-SEAL thread, and tighten cap onto thread to make a vacuum seal. Use with 5mm × 9in. L NMR tubes.
Leistungsmerkmale und Vorteile
  • Manufactured from borosilicate glass 3.3
  • NMR sealing manifolds utilize KONTES® HI-VAC valves
  • NMR manifolds feature a BEVEL-SEAL threaded connection and a specially designed cap with an aluminum shield for heat deflection
  • All O-Rings supplied are FKM
  • Valve plugs are 826501-0008
Rechtliche Hinweise
BEVEL-SEAL is a trademark of Kimble Kontes Asset Mgmt. Inc.
HI-VAC is a registered trademark of Kimble Kontes Asset Mgmt. Inc.
KIMBLE is a registered trademark of DWK Life Sciences
Kontes is a registered trademark of Kimble Kontes Asset Mgmt. Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z255696-1EA4061832901596