Z218898
Ace giant extraction thimble
I.D. × L 60 mm × 180 mm
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Empfohlene Produkte
Hersteller/Markenname
Ace Glass 681202
ID × L
60 mm × 180 mm
Allgemeine Beschreibung
For use with Ace giant Soxhlet extractors. Thimbles are readily permeable to the flow of ether and other organic solvents. Made of cellulose.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)