Merck
Z214558
Micro photochemical reaction assembly
with quartz well
About This Item
UNSPSC-Code:
41121800
Leistungsmerkmale
CE compliant: no
CSA approved: no
Hersteller/Markenname
Ace Glass 788060
Verfügbarkeit
not available in EU, Canada
Allgemeine Beschreibung
New reactor design1 handles volumes from 5 to 15 mL. Borosilicate glass body with #25 Ace-Thred at top, 14/10 outer side joint externally threaded to allow an O-ring cap-seal connection with mating male joints, and a porosity E (4 to 8 μm) filter disc for use as a gas inlet to remove oxygen from the solution or for agitation. Includes an 11 mm I.D. jacketed quartz immersion well, and a quartz Pen-Ray 5.5 W, low pressure, cold cathode, mercury lamp with matching 115 V, 60 Hz power supply.
1.Designed by John H. Penn and Richard D. Orr, West Virginia Univ. Dept. of Chemistry, Morgantown, WV 26506.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z214558-1EA4061833292723