Merck
SYNV190390
Synthware Double-Walled Quartz Photochemical Immersion Well,
L 390 mm
Synonym(e):
Synthware V190390, immersion well
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.51
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware V190390
Länge
390 mm
Verwandte Kategorien
Allgemeine Beschreibung
Double-walled quartz immersion well for use with V150500 reactor and lamp. A small diameter inner tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet. Well is used with RC50 Nylon Bushing with Viton® O-ring to form a compression type seal with threaded vessels. Well offers a greaseless connection and can be adjusted to desired height .The lack of the ground joint means easier dismantling since there is no freezing.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Ähnliches Produkt
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)