Merck
Z187224
Sigma-Aldrich
Lecture-bottle control valve with compression fitting
tube outlet O.D. 1/4 in., stainless steel
Synonym(e):
Lecture Bottle Control Valve
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Materialien
stainless steel
Röhrchenausgang AD
1/4 in.
Allgemeine Beschreibung
CGA 110 requires CGA 110 PTFE sealing washer.
Warnhinweis
WARNING: Gas-control valves do not control pressure in a closed system. Regulator or relief valves should be used to prevent the buildup of unsafe pressures that could lead to explosion.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)