Z155969
Edelstahl-Trockenwannen
rolled bead, L × W × H 15.1 in. × 10.6 in. × 0.75 in.
Alle Fotos(1)
About This Item
UNSPSC-Code:
41104510
Empfohlene Produkte
Hersteller/Markenname
Polarware 15F
L × B × H
15.1 in. × 10.6 in. × 0.75 in.
Allgemeine Beschreibung
Stainless steel drying trays have a low profile, and are lighter weight than glass. We carry a wide range of sizes. The unbreakable stainless steel material has rapid cooling and heating for your samples.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)