WIMWG12091
Multi-tube jet washer/dryer, 5-position, for 3-5 mm NMR tubes
for #9 silicone joint
Synonym(e):
cleaning nmr tubes, tube washers
Alle Fotos(1)
About This Item
UNSPSC-Code:
47131800
NACRES:
NB.43
Empfohlene Produkte
Verpackung
pkg of 1 ea
Hersteller/Markenname
Wilmad WG-1209-1
Fitting
for #9 silicone joint
Allgemeine Beschreibung
This Wilmad Multi-Tube Jet Solvent Washer/Dryer is recommended for research labs that routinely clean NMR and EPR tubes; a single unit can accommodate up to 5 tubes (3, 4 and 5 mm OD) at once. When an inverted tube is inserted onto the solvent transfer tubing and the open end is immersed under wash solvent in the solvent cup, a reliable vacuum-tight seal will be formed and generate solvent flow under vacuum. After solvent is fully consumed, air flow will follow to turn the unit into a dryer.
Features:
Features:
- 5 PTFE coated stainless steel solvent transfer tubes fit 3, 4 and 5 mm NMR/EPR tubes
- PTFE solvent cup and tubing make this unit fully resistant to common organic solvents
- Flanged reservoir connection eliminates joint freeze
- Greaseless joint between the solvent cup and glass reservoir eliminates possibility of contamination
- Complete disassembly without tools for easy cleaning
- Hands free during washing/drying cycle
- Calibrated length mark for 4 in., 7 in., 8 in. and 9 in. tube
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)