Merck
WHAMUPMCBT8
Whatman® Mini-UniPrep® syringeless filters accessories
Multicompressor tray
About This Item
UNSPSC-Code:
41104922
NACRES:
NB.24
Leistungsmerkmale
Multicompressor tray
Hersteller/Markenname
Whatman MUPMCBT8
Allgemeine Beschreibung
Single 8 position blue tray to be used with Plastic MUP
Leistungsmerkmale und Vorteile
  • Designed for the new Whatman Mini-UniPrep8-vial Multicompressor
  • Holds eight Mini-UniPrepstandard plastic filter vials for simultaneous processing
  • Get a spare tray for added convenience
Rechtliche Hinweise
Uniprep is a registered trademark of Whatman International Ltd.
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)