Merck
WHA10350240
Whatman® Extraktionshülsen aus Cellulose
603 Cellulose thimbles, 33 x 80 mm - thickness 1.5 mm, 25/pk
Synonym(e):
Z696501, cellulose thimble, extraction thimble, glass thimbles
About This Item
UNSPSC-Code:
41104817
NACRES:
NB.43
Verpackung
pack of 25
Hersteller/Markenname
Whatman 10350240
Whatman Article No. 28414697 (US reference)
B × L
33 mm × 80 mm
Dicke
1.5 mm
Leistungsmerkmale und Vorteile
  • 603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes.
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)