TEM-LC200AUUL
Ultrathin Lacey Carbon Supported Gold Grid
size 200 mesh, box of 25
Synonym(e):
LACEY CARBON FILM - AU 200 MESH ULTRA THIN 25/BX
Alle Fotos(1)
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Empfohlene Produkte
Verpackung
box of 25
Größe
200 mesh
Verwandte Kategorien
Allgemeine Beschreibung
Ultrathin Lacey Carbon (4-6 nm) Supported Grid: Gold Grid, 200 mesh. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)