Merck
TEM-FFT200CUUL
Sigma-Aldrich
Ultrathin Formvar Supported Copper Thin Bar Square Mesh
size 200 mesh, box of 50
Synonym(e):
FORMVAR THIN BAR SQUARE MESH - CU 200 MESH UL 50/BX
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Verpackung
box of 50
Größe
200 mesh
Allgemeine Beschreibung
A 5-6 nm thin film of pure formvar resin supported thin bar square mesh grids. Copper grid, 200 mesh.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)