Merck
TEM-FF100CUUL
Sigma-Aldrich
Ultrathin Formvar Supported Copper Square Mesh
size 100 mesh, box of 50
Synonym(e):
FORMVAR SQUARE MESH - CU 100 MESH UL 50/BX
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Verpackung
box of 50
Größe
100 mesh
Allgemeine Beschreibung
A 5-6 nm ultrathin formvar film supported standard copper square mesh grids. Copper grid, 100 mesh. All of our film is laid on the shiny side of the grid.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)