Merck
SYNF561025
Synthware reaction tube with high vacuum valve
25 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonym(e):
Synthware F561025, Schlenk flask
About This Item
UNSPSC-Code:
41121700
NACRES:
NB.51
Materialien
colorless tube
glass tube
Verbindung
joint: ST/NS14/20
Sterilität
non-sterile
Leistungsmerkmale
graduations: no
Push fit
Verpackung
pkg of 1 ea
Hersteller/Markenname
Kemtech F561025
Kapazität
25 mL
25 mL
Ventilgröße
0-4 mm
Allgemeine Beschreibung
Supplied with high vacuum valve with PTFE o-ring at tip and PTFE wiper (“umbrella”) to protect Viton® O-ring on shaft.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)