Merck
TEM-CF300HCU50
Sigma-Aldrich
Carbon Film Supported Copper Hexagonal Mesh Grid
size 300 mesh, box of 50
Synonym(e):
CARBON FILM ONLY ON 300 HEX COPPER GRIDS 50/BX
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Materialien
carbon film
copper grid
Beschreibung
film layer thickness 5 - 6 nm carbon
Verpackung
box of 50
Größe
300 mesh
Allgemeine Beschreibung
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 300 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)