Merck
TEM-CF400-CU-TH
Sigma-Aldrich
Carbon Film Supported Copper Square Mesh
Thick, size 400 mesh, box of 50
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Verpackung
box of 50
Größe
400 mesh
Allgemeine Beschreibung
A 10 nm thick film of pure carbon supported standard squre mesh grids. Copper grid, 400 mesh.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)