SYNS560008
Synthware™ 180 degree high vacuum valve
size 0-8 mm
Synonym(e):
Synthware S560008
Alle Fotos(1)
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
Empfohlene Produkte
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware S560008
Allgemeine Beschreibung
With two 9 mm O.D. sidearms sealed to the barrel at a 90° angle to one another. The non-rotating PTFE shaft has three O-rings, the tip O-ring making the primary seal on the precision formed tapered seat . For replacement Viton® O-ring Kit, see R7010,or R108110.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)