Merck
SYNP120010M
Synthware peptide synthesis vessel with fritted disc and PTFE stopcock
10 mL, disc O.D. 15 mm, porosity: medium, GL 14
Synonym(e):
Synthware P120010M, Solid phase peptide synthesis vessel
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.51
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware P120010M
Kapazität
10 mL
10 mL
Scheibe AD
15 mm
Fitting
GL 14
Allgemeine Beschreibung
With a fritted disc of coarse or medium porosity to support resin. Top of the vessel has GL thread and a PTFE lined screw cap. PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)